missing translation for 'onlineSavingsMsg'
Learn More

Aluminum hydroxide, 76.5% min

Product Code. 11311759
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11311759

Brand: Thermo Scientific Alfa Aesar 012366.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol

As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmeticsAluminum hydroxide is used as a desiccant powder, filler in paper, plastics, rubber and cosmetics. It is used as a smoke suppressant and mordant dye. It is also used in drugs as antacid and antihyperphosphatemic. It is also used as a Claus catalyst support for waterproof fabrics. It is an important starting material for the preparation of other aluminum compounds, calcined alumina, aluminum sulfate, polyaluminum chloride, zeolites, sodium aluminate, activated alumina and aluminum nitrate. In addition, it is used as a fire retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]

Specifications

Physical Form Granules
Quantity 100 g
Assay Percent Range 76.5% min.
Solubility Information Soluble in alkaline aqueous solution,hydrochloric acid and sulfuric acid solution. Insoluble in water.
Formula Weight 78
Grade Reagent
Chemical Name or Material Aluminum hydroxide
compliance-icons
Product Identifier
  • Aluminum hydroxide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.