missing translation for 'onlineSavingsMsg'
Learn More

Aluminum hydroxide, extra pure, powder, Thermo Scientific Chemicals

£24.22 - £234.17

Chemical Identifiers

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10482791
View Documents
Thermo Scientific Acros
219130250
25 g Plastic bottle
£24.22
25g
Please to purchase this item. Need a web account? Register with us today!
10752261
View Documents
Thermo Scientific Acros
219135000
500 g Plastic bottle
£28.34
500g
Please to purchase this item. Need a web account? Register with us today!
10328070
View Documents
Thermo Scientific Acros
219130020
2 kg Plastic bottle
£71.05
2kg
Please to purchase this item. Need a web account? Register with us today!
11944141
View Documents
Thermo Scientific Acros
219130100
10 kg Plastic drum
£234.17
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21645-51-2
78.00
WNROFYMDJYEPJX-UHFFFAOYSA-K
[OH-].[OH-].[OH-].[Al+3]
AlH3O3
MFCD00003420
aluminium(3+) trihydroxide

Specifications

21645-51-2
8.5 to 10 (5% aq. suspension)
AlH3O3
MFCD00003420
Solubility in water: insoluble
[OH-].[OH-].[OH-].[Al+3]
78.00
≥63.5% (Al2O3)
36.5% max.
White
Powder
Al(OH)3
15,338
WNROFYMDJYEPJX-UHFFFAOYSA-K
aluminium(3+) trihydroxide
78
Extra Pure
Aluminum hydroxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 244-492-7

Product Identifier
  • Aluminium hydroxide

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.