Learn More
Aluminum iodide, 95%, Thermo Scientific Chemicals
Description
Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 360°C |
Melting Point | 191°C |
Physical Form | Chunks |
Quantity | 25 g |
Assay Percent Range | 95% |
Solubility Information | Soluble in alcohol,ether,carbon disulfide |
Formula Weight | 407.69 |
Sensitivity | Light and moisture sensitive |
Density | 3.98 |
Chemical Name or Material | Aluminum iodide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M22L020 | Certificate of Analysis | 24/01/2025 | 030714.22,030714.14 |
Y22I042 | Certificate of Analysis | 17/01/2023 | 030714.22,030714.14 |
G08X013 | Certificate of Analysis | 08/12/2021 | 030714.22,030714.14 |
N11B027 | Certificate of Analysis | 08/12/2021 | 030714.22,030714.14 |
W28H019 | Certificate of Analysis | 08/12/2021 | 030714.22,030714.14 |
Safety and Handling

- Aluminum iodide
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.