Learn More
Brady™ Aluminum: ISO Safety Sign - Safety shower
Sign compliant to ISO 7010.
Brand:  Brady™ 815723
Additional Details : Weight : 0.00500kg
 View more versions of this product
Product Code. 15674347
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Aluminum
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
1 Ea.
100 mm
100 mm
Square
White on Green
Aluminum
Indoor and Outdoor
1.016 mm
-40°C to 140°C
Global