missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum: ISO Safety Sign - Warning; Automatic start-up

Sign compliant to ISO 7010.

Brand:  Brady™ 828990

Additional Details : Weight : 0.01300kg

 View more versions of this product

Product Code. 15684747

  • £6.15 / Each
Estimated Shipment: 21-08-2024
to see stock.
Add to basket

Description

Description

  • Aluminum
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
1 Ea.
87 mm
100 mm
Triangular
Black on Yellow
Aluminum
Indoor and Outdoor
1.016 mm
-40°C to 140°C
Global
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers