Learn More
Welch™ Aluminum KF Inner Centering with Viton O-ring
Designed for use with CRVpro Rotary Vane Vacuum Pumps. Welch Ilmvac™ Aluminum KF Inner Centering Rings feature a replaceable Fluoroelastomer O-ring seal.
Brand: Welch™ 701095
This item is not returnable.
View return policy
Description
- Stainless steel centering ring with Viton O-ring
- Replaceable Fluoroelastomer O-ring seal
- Centering ring assemblies are placed between two ISO flanges with matching outer diameters
- he widest portion of the centering ring rests inside a centering ring locater groove (blank flange) or counterbore (bored flange) on the Kwik-Flange™
- The O-ring rests on the flat polished surface outside the locater groove or counterbore
Specifications
Separator 40 DN | |
Aluminum | |
1 Each | |
KF Inner Centering |
Product Content Correction
Product Title
Welch™ Aluminum KF Inner Centering with Viton O-ring > O.D.: 40mm
Spot an opportunity for improvement?Share a Content Correction