missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Aluminum Light Crucible

Brand:  Mettler Toledo™ 51119810

Code : X3

Additional Details : Weight : 0.10000kg

Product Code. 12744942

  • £436.74 / Each
Estimated Shipment: 03-10-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Aluminum
1Pack
1.6mm
640°C
20 μL
Light Crucible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers