missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009 in.) dia. wire, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 040906.HB
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Aluminum Magnesium gauze, Alloy 5056 | |
Al:Mg; 95:5 wt% | |
300 x 300 mm | |
Odorless |
Woven from 0.23mm (0.009 in.) dia. wire | |
MFCD00214039 | |
30 Mesh | |
Wire Cloth |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009 in.) dia. wire, Thermo Scientific Chemicals > 300x300mm
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction