missing translation for 'onlineSavingsMsg'
Learn More
Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals Product Code.: 11393728

Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals

Product Code. 11393728
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11393728

Brand: Thermo Scientific Alfa Aesar 039320.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69207-83-6
MDL Number MFCD00070426

Specifications

Melting Point 40°C
Boiling Point 114°C (0.2 mmHg)
Quantity 10 g
UN Number UN3181
Sensitivity Moisture sensitive
Formula Weight 550.53
Percent Purity ≥99.95%
Physical Form Crystalline Low Melting Solid
Assay (metals basis)
Chemical Name or Material Aluminum magnesium isopropoxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN18F012 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number039320.04,039320.18,039320.09
Lot NumberB15W013 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number039320.04,039320.18,039320.09
Lot NumberN18F012 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number039320.04,039320.18,039320.09
Lot NumberH28L11 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number039320.04,039320.18,039320.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.