missing translation for 'onlineSavingsMsg'
Learn More

Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%, Thermo Scientific Chemicals

Product Code. 11387328
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum metaphosphate is used as a constituent of glazes, enamels, glasses and as a high-temperature insulating cement. It is also used as molecular sieves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32823-06-6
MDL Number MFCD00075314

Specifications

Color White
Physical Form Powder
Quantity 5 x 1 kg
Solubility Information Insoluble in water.
Formula Weight 263.9
Concentration or Composition (by Analyte or Components) Al2O3 19.4%, P2O5 78.9%
Density 2.779 g/mL
Chemical Name or Material Aluminum metaphosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'