Learn More
Aluminum n-butoxide, 90+%, Thermo Scientific Chemicals
£114.00 - £429.00
Chemical Identifiers
CAS | 3085-30-1 |
---|---|
Molecular Formula | C12H30AlO3 |
Molecular Weight (g/mol) | 249.351 |
MDL Number | MFCD00015237 |
InChI Key | YGVBJFIUURLJDC-UHFFFAOYSA-N |
Synonym | aluminum iii n-butoxide, aluminum; butan-1-ol |
PubChem CID | 50933522 |
IUPAC Name | aluminum;butan-1-ol |
SMILES | CCCCO.CCCCO.CCCCO.[Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355029
|
Thermo Scientific Alfa Aesar
087754.09 |
10 g |
£114.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345029
|
Thermo Scientific Alfa Aesar
087754.18 |
50 g |
£429.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3085-30-1 | |
249.351 | |
YGVBJFIUURLJDC-UHFFFAOYSA-N | |
50933522 | |
CCCCO.CCCCO.CCCCO.[Al] |
C12H30AlO3 | |
MFCD00015237 | |
aluminum iii n-butoxide, aluminum; butan-1-ol | |
aluminum;butan-1-ol |
Specifications
102°C | |
Lump | |
Al[O(CH2)3CH3]3 | |
UN3181 | |
YGVBJFIUURLJDC-UHFFFAOYSA-N | |
aluminum;butan-1-ol | |
50933522 | |
≥90% | |
Aluminum n-butoxide |
3085-30-1 | |
C12H30AlO3 | |
MFCD00015237 | |
aluminum iii n-butoxide, aluminum; butan-1-ol | |
CCCCO.CCCCO.CCCCO.[Al] | |
249.351 | |
246.32 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 221-394-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only