missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitride, N 32.0% min

Product Code. 11377247
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
5g
This item is not returnable. View return policy

Product Code. 11377247

Brand: Thermo Scientific Alfa Aesar 011546.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24304-00-5 | AlN | 40.99 g/mol

Aluminum nitride possesses very high thermal conductivity and effectively used in the ceramic industry. It finds application in deep ultraviolet optoelectronics, steel production, dielectric layers in optical storage media, chip carriers and as a crucible to grow crystals of gallium arsenide. It is also used as a radio frequency filter, which finds application in mobile phones. Further, it is used as a circuit carrier in semiconductors. In addition to this, it is used as a heat-sink in light-emitting diode lighting technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24304-00-5
Molecular Formula AlN
Molecular Weight (g/mol) 40.99
MDL Number MFCD00003429
InChI Key PIGFYZPCRLYGLF-UHFFFAOYSA-N
Synonym aluminum nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminum nitride grade a, aluminum nitride grade b, aluminum nitride grade c
PubChem CID 90455
ChEBI CHEBI:50884
SMILES N#[Al]

Specifications

Melting Point >2200°C
Color White
Physical Form Powder
Quantity 50 g
Merck Index 14,353
Solubility Information Soluble in mineral acids.
IUPAC Name alumanylidyneamine
Formula Weight 40.99
Concentration or Composition (by Analyte or Components) N 32.0% min.
Odor Ammonia-like
Sensitivity Moisture sensitive
Density 3.26 g/mL
Chemical Name or Material Aluminum nitride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.