missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum with Over laminated Polyester Transport Signs
Brand: Brady™ 813601
Code : NEW
Additional Details : Weight : 0.24300kg
Specifications
ISO 7010 | |
1 Ea. | |
Used for indoor and outdoor safety signs | |
297 mm | |
Rhomboid | |
Transport Sign |
Aluminum, Polyester | |
Aluminum with Polyester Overlaminate | |
0.271 mm | |
-40°C to 140°C | |
Environmentally Hazardous Substances | |
Black on White |