missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxalate hydrate, 99%

Product Code. 11379648
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11379648

Brand: Thermo Scientific Alfa Aesar 044149.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 814-87-9 | C6Al2O12 | 318.02 g/mol

Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-87-9
Molecular Formula C6Al2O12
Molecular Weight (g/mol) 318.02
MDL Number MFCD00050791
InChI Key ZCLVNIZJEKLGFA-UHFFFAOYSA-H
Synonym aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate
PubChem CID 93561
IUPAC Name dialuminum;oxalate
SMILES [Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Color White
Linear Formula Al2(C2O4)3·xH2O
Quantity 1 kg
Merck Index 14,355
Solubility Information Insoluble in water.
Formula Weight 318.02 (Anhydrous)
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material Aluminum oxalate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.