missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Aluminum Oxide 90 Acidic
Brand: Macherey-Nagel™ 815030.5
This item is not returnable.
View return policy
Specifications
50 to 200 μm | |
4 ± 0.3 | |
∼130 m2/g |
Activity | |
5 kg | |
Aluminum Oxide 90 Acidic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Aluminum Oxide 90 Acidic > Quantity: 5kg
Spot an opportunity for improvement?Share a Content Correction