missing translation for 'onlineSavingsMsg'
Learn More
Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals Product Code.: 11327187

Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals

Product Code. 11327187
5 kg
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11327187

Brand: Thermo Scientific Alfa Aesar 011502.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Neutral; S.A. 150m2/g

Aluminum oxide is used as an abrasive, dosimeter, filler of choice for plastics, ingredient in sunscreen and cosmetics. Aluminum oxide flakes are also used in paint for providing reflective decorative effects. In laboratories it is widely used as a medium for chromatographic purifications. It is used in the production of aluminum (Hall–Héroult process), alumina nanofibers, cutoff tools, and sandpaper. It serves as a catalyst for several reactions of industrial importance, including converting alcohols into alkenes, decomposing hydrazine, drying of gases, and converting hydrogen sulfide into sulfur in refineries (Claus Process). It is also used in integrated circuits, compact fluorescent lamps (CFL), sodium vapor lamps, hip replacements, CD/DVD polishing, superconducting quantum interference devices (SQUIDS), insulation devices, and as a support for many popular catalysts (for example, in Ziegler-Natta polymerizations).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Melting Point 2045°C
Physical Form Powder
Quantity 5 kg
Solubility Information Insoluble in water,ethanol and diethyl ether.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade Brockmann
Odor Odorless
Appearance White powder
Chemical Name or Material Aluminum oxide, activated, neutral, 58 angstroms

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.