missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide, Basic, Brockmann Activity Grade I, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Descripción
- For Chromatography
Especificaciones
Especificaciones
Physical Form | Powder |
Quantity | 500 g |
IUPAC Name | dialuminum;oxygen(2-) |
Formula Weight | 101.96 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 4.00kg |
Chemical Name or Material | Aluminum Oxide, Basic, Brockmann Activity Grade I |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'