missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, fused, insulating powder, 99.7+%, Thermo Scientific Chemicals

Used in Dry Filling For thermal Insulation of Electrical Furnaces

£63.92 - £235.17

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313138
View Documents
Thermo Scientific Alfa Aesar
033200.A1
1 kg
£63.92
1kg
Please to purchase this item. Need a web account? Register with us today!
11303138
View Documents
Thermo Scientific Alfa Aesar
033200.A7
5 kg
£235.17
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in dry filling for thermal insulation of electrical furnaces.Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fiber. Heavy quantity of aluminum oxide is used in the production of aluminum and a component of clays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Form: approx. 60 micron
Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)

Specifications

1344-28-1
Al2O3
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
White powder
Aluminum oxide
>99.7%
MFCD00003424
Insoluble in water,but slightly soluble in acid and alkali.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
Odorless
1.33 hPa (1mmHg) at 2158°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

missing translation for 'rtecsNumber' : BD1200000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only