Learn More
Aluminum oxide, Gamma phase, Catalyst support, High surface area, Bimodal, Thermo Scientific Chemicals
Description
Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 2980°C |
Melting Point | 2045°C |
Physical Form | Pellets |
Quantity | 5 kg |
Solubility Information | Insoluble in water. Slightly soluble in acid and alkali. |
IUPAC Name | dialuminum;oxygen(2-) |
Formula Weight | 101.96 |
Density | 3.965 |
Chemical Name or Material | Aluminum oxide, Catalyst Support |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z09K030 | Certificate of Analysis | 11/12/2024 | 043832.A1,043832.30,043832.18,043832.A7 |
W12J027 | Certificate of Analysis | 21/09/2023 | 043832.A1,043832.30,043832.18,043832.A7 |
W21I033 | Certificate of Analysis | 13/10/2022 | 043832.A1,043832.30,043832.18,043832.A7 |
L02T031 | Certificate of Analysis | 08/12/2021 | 043832.A1,043832.30,043832.18,043832.A7 |
L03N09 | Certificate of Analysis | 08/12/2021 | 043832.A1,043832.30,043832.18,043832.A7 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.