missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide-silicon oxide (13%), catalyst support, low surface area, macroporous, Thermo Scientific Chemicals
Form: S.A. 0.008m2/g
£169.85 - £736.67
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
IUPAC Name | dialuminium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11390538
|
Thermo Scientific Alfa Aesar
043863.A1 |
1 kg |
£169.85
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11300548
|
Thermo Scientific Alfa Aesar
043863.A7 |
5 kg |
£736.67
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum oxide-silicon oxide is a fibrous material used as loose wool, blanket, felt, paper or boards.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
dialuminium(3+) trioxidandiide |
Specifications
1344-28-1 | |
Spheres | |
MFCD00003424 | |
Insoluble in water. | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
Odorless | |
Aluminum oxide |
White | |
Al2O3 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
White spheres |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only