missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, single crystal, Thermo Scientific™

Product Code. 15431467
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11092-32-3
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key TWNQGVIAIRXVLR-UHFFFAOYSA-N
Synonym alumina, corundum, electrocorundum, korund, emery, alpha-corundum, abramant f 36, abramax f 46, abramax f 54
PubChem CID 14769
IUPAC Name oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O

Specifications

Quantity 1 Pc.
Solubility Information Insoluble in water.
Chemical Name or Material Aluminum oxide, Single crystal, 25.4mm dia x 0.5mm thick, random orientation
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'