Learn More
Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml, Thermo Scientific Chemicals
£37.99 - £135.68
Chemical Identifiers
CAS | 7784-27-2, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | AlH18N3O18 |
Molecular Weight (g/mol) | 375.129 |
MDL Number | MFCD00134029 |
InChI Key | SWCIQHXIXUMHKA-UHFFFAOYSA-N |
PubChem CID | 24567 |
IUPAC Name | aluminum;trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309017
|
Thermo Scientific Alfa Aesar
038727.AD |
50 mL |
£37.99
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399007
|
Thermo Scientific Alfa Aesar
038727.AE |
100 mL |
£55.48
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400181
|
Thermo Scientific Alfa Aesar
038727.AP |
500 mL |
£135.68
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-27-2, 7697-37-2, 7732-18-5 | |
375.129 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate |
AlH18N3O18 | |
MFCD00134029 | |
24567 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
Specifications
7784-27-2, 7697-37-2, 7732-18-5 | |
AlH18N3O18 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 | |
23 hPa (17mm Hg) at 20°C | |
Aluminum, plasma standard solution, Al 1000μg/mL |
Colorless | |
MFCD00134029 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.129 | |
Nitric acid: 5.0%; Aluminum nitrate nonahydrate: 1.4%; Water: 93.6% | |
Liquid |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 231-714-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum, plasma standard solution, Specpure Al 1000µg/ml
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.