missing translation for 'onlineSavingsMsg'
Learn More
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals Product Code.: 11414283

Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Product Code. 11414283
500 g
Click to view available options
Quantity:
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11414283

Brand: Thermo Scientific Alfa Aesar A10906.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.372
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrateShow MoreShow Less
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Melting Point 92°C
Quantity 500 g
Merck Index 14,360
Formula Weight 474.38 (258.20 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Density 1.757
Chemical Name or Material Aluminum potassium sulfate dodecahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250975 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA10906.0E,A10906.36
Lot Number10245483 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog NumberA10906.0E,A10906.36
Lot Number10196168 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA10906.0E,A10906.36
Lot Number10121334 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10906.0E,A10906.36
Lot Number10208141 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10906.0E,A10906.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.