missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1 kg
missing translation for 'unitSize'
1kg
250g
50g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Color | Silver to Gray |
| Appearance | Silver Gray powder |
| Quantity | 250 g |
| Ignition Point | 400°C (752°F) |
| Mesh Size | -100+325 Mesh |
| Odor | Odorless |
| Sensitivity | Water reactive |
| Assay Percent Range | 99.5% (Metals basis) |
| Form | Powder |
| IUPAC Name | aluminium |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Aluminum powder
- Danger
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P370+P378-In case of fire: Use to extinguish.
- P422-Store contents under .
- P501-Dispose of contents/container to .
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur