missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Silver to Gray |
Appearance | Silver Gray powder |
Density | 2.7g/cm3 |
Assay Percent Range | 99.5% (Metals basis) |
Quantity | 250 g |
Ignition Point | 400°C (752°F) |
IUPAC Name | aluminium |
Mesh Size | -325 Mesh |
Odor | Odorless |
Sensitivity | Water reactive |
Show More |
Safety and Handling
Product Identifier
- Aluminum powder
- Danger
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P232-Protect from moisture.
- P370+P378-In case of fire: Use fire-fighting equipment on basis class D to extinguish.
- P402-Store in a dry place.
- MIXTURE LIST-Contains : Aluminium
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction