missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Frame Rod Platform, Aluminum

Use the Frame Rod Platform to provide support for your specific laboratory equipment.

Brand:  Cole-Parmer™ Essentials 08059-30

Product Code. 11797583

  • £362.00 / Each
Estimated Shipment: 30-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Use to support your labware items from lab fames or support stands.
  • The round plate design features a raised rim to prevent your apparatus from falling.
    • Requires an additional connector in order to attach clamps to frames or other apparatus.
  • The rectangular plates include an integral connector.
Specifications

Specifications

Aluminum
15.75 ft.
12 ft.
Rectangular Support Plate
For Specific Laboratory Equipment
15.75 x 12 ft.
400 mm
304.8 mm
Rectangular
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Essentials Frame Rod Platform, Aluminum > Frame Rod Platform; Material: Aluminum; Dimensions (L x W): 15.75 x 12 ft.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.