missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Aluminum Rods

Product Code. 10711611
Click to view available options
Length (Metric):
500 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10711611

Brand: Bochem™ 5130

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Stand Bases M10 Threaded

  • Aluminum
  • With thread
  • Sizes vary
  • For stand base
  • 12 D/O

Specifications

Material Aluminum
Length (Metric) 500 mm
Length (Metric) Rod 500 mm
Diameter (Metric) Outer 12 mm
Product Type Rod
Thread Size M10
For Use With (Application) For Stand Bases M10 Threaded
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.