missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Teflon FEP Coated Aluminum Scoops

Teflon FEP Coated Aluminum Scoops are FDA compliant for food contact

Brand:  Bel-Art™ SP Scienceware™ H36735-0000

 View more versions of this product

Product Code. 11741495

  • £130.12 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Anti-stick, corrosion resistant brown coating
  • Complies with FDA regulations for food contact
  • Handle is not coated so your grip is non-slip
Specifications

Specifications

Scoop
Sampling Scoop
0.5 gal.
Brown
FDA Compliant for Food Contact
311 x 127 x 89 mm
89 mm
209 mm
127 mm
Aluminum
Non-Slip Grip
2 L
Teflon™
311 x 127 mm
89 mm
311 mm
102 mm
127 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers