Learn More
Corning™ Aluminum Sealing Film for General and Thermal Sealing
Provides exceptional seal on multi-well plates with varied temperatures and reagents. Corning™ Aluminum Sealing Film for General and Thermal Sealing is designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture.
Brand: Corning™ PCR-AS-600
116.85 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : A0
Additional Details : Weight : 0.27900kg
Description
- Seals during thermocycling when used in conjunction with compression mats
- Ideal for light-sensitive samples
- Uniformly applied adhesive provides optimum well sealing
Specifications
Aluminum sealing film | |
Non-sterile | |
146 x 79.2 mm | |
35 μm |