missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sec-butoxide, 97%

Product Code. 11409983
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11409983

Brand: Thermo Scientific Alfa Aesar A13044.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2269-22-9 | C12H30AlO3 | 249.351 g/mol

Aluminum sec-butoxide is used to prepare photosensitive alumina gel films. It is also used in the preparation of microporous crystalline aluminophosphate AlPO-41 and alumina mesoporous material. It is used as a gelling agent of paint and ink and water repellent substance of wood. Further, it acts as an active catalyst and reducer of aldehydes and ketones. In addition to this, it is used to make high-surface-area alumina using 2,4-pentanedione as a chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum sec-butoxide is used to prepare photosensitive alumina gel films. It is also used in the preparation of microporous crystalline aluminophosphate AlPO-41 and alumina mesoporous material. It is used as a gelling agent of paint and ink and water repellent substance of wood. Further, it acts as an active catalyst and reducer of aldehydes and ketones. In addition to this, it is used to make high-surface-area alumina using 2,4-pentanedione as a chelating agent.

Solubility
Miscible with alcohol, isopropyl alcohol and toluene.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, metals and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2269-22-9
Molecular Formula C12H30AlO3
Molecular Weight (g/mol) 249.351
MDL Number MFCD00009327
InChI Key LWGPRERTOLVRLK-UHFFFAOYSA-N
Synonym aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide
PubChem CID 50931103
IUPAC Name aluminum;butan-2-ol
SMILES CCC(C)O.CCC(C)O.CCC(C)O.[Al]

Specifications

Boiling Point 200°C to 206°C (30mmHg)
Quantity 100 g
UN Number UN1992
Beilstein 3910908
Flash Point 27°C (82°F)
Solubility Information Miscible with alcohol,isopropyl alcohol and toluene.
Formula Weight 246.32
Percent Purity 97%
Odor Alcohol-like
Refractive Index 1.439
Sensitivity Moisture sensitive
Density 0.967
Chemical Name or Material Aluminum sec-butoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.