missing translation for 'onlineSavingsMsg'
Learn More

Aluminum selenide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11362228
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a precursor to hydrogen selenide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-82-5
MDL Number MFCD00014158
Synonym unii-4r7pmp996q, aluminum selenide, aluminum selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide

Specifications

Color Brown
Physical Form Pieces
Quantity 1 g
UN Number UN3283
Merck Index 14,361
Solubility Information Decomposes to Al203 and SeO2 in water
Formula Weight 290.84
Percent Purity 99%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.437 g/mL
Assay (metals basis)
Chemical Name or Material Aluminum selenide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aluminum selenide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Precautionary Statement
  • P301-IF SWALLOWED:.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P330-Rinse mouth.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P373-DO NOT fight fire when fire reaches explosives.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P410-Protect from sunlight.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'