missing translation for 'onlineSavingsMsg'
Learn More
Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11349927

Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11349927
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11349927

Brand: Thermo Scientific Alfa Aesar 042258.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 50 g
Percent Purity 99.99%
Odor Odorless
Weight ≈0.14g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ10K009 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog Number042258.30,042258.18
Lot NumberZ30H012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042258.09,042258.30,042258.18
Lot NumberU21F043 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number042258.09,042258.30,042258.18
Lot NumberN06F012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number042258.09,042258.30,042258.18
Lot NumberE01L01 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042258.09,042258.30,042258.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.