missing translation for 'onlineSavingsMsg'
Learn More

Aluminum spheres, 2.38mm (0.094in) dia, 99% (metals basis), Thermo Scientific Chemicals

£39.20 - £106.00

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11395959
View Documents
Thermo Scientific Alfa Aesar
042029.KM
25 Pc.
£39.20
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
11305969
View Documents
Thermo Scientific Alfa Aesar
042029.KN
100 Pc.
£106.00
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629
[Al]

Specifications

7429-90-5
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
(metals basis)
Alloy 1100, Al:Cu; ≥99:0.12 wt%
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
5359268
99%
Aluminum spheres, 2.38mm (0.094 in.) dia.
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-072-3

missing translation for 'rtecsNumber' : BD0330000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only