missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.999% (metals basis), Thermo Scientific™
£425.00 - £1718.00
Chemical Identifiers
CAS | 7429-90-5 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
IUPAC Name | aluminum |
SMILES | [Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11305117
|
Thermo Scientific Alfa Aesar
040829.KS |
1 Ea. |
£425.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395107
|
Thermo Scientific Alfa Aesar
040829.KU |
5 Ea. | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
aluminum |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
CHEBI:33629 | |
[Al] |
Specifications
7429-90-5 | |
MFCD00134029 | |
Insoluble in water. | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Odorless | |
Aluminum sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick |
Al | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminum | |
5359268 | |
99.999% | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 231-072-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures