missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£89.60 - £226.00
Chemical Identifiers
CAS | 7429-90-5, 7732-18-5, 7697-37-2 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
IUPAC Name | aluminium |
SMILES | [Al] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11428619
|
SPEX Certiprep
PLAL2-2Y |
125 mL | Triple-leached LDPE bottle |
£89.60
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10446572
|
SPEX Certiprep
PLAL2-2T |
250 mL | Triple-leached LDPE bottle |
£127.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041684
|
SPEX Certiprep
PLAL2-2X |
500 mL | Triple-leached LDPE bottle |
£226.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Al | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7429-90-5, 7732-18-5, 7697-37-2 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
CHEBI:33629 | |
[Al] |
Al | |
MFCD00134029 | |
5359268 | |
aluminium |
Specifications
7429-90-5, 7732-18-5, 7697-37-2 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00134029 | |
[Al] | |
26.98 | |
CHEBI:33629 | |
2% HNO3 |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Al | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminium | |
5359268 | |
Assurance | |
Aluminum Standard |
Safety and Handling
Causes skin irritation, serious eye irritation
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only