missing translation for 'onlineSavingsMsg'
Learn More

Aluminum tert-butoxide, 97%, Thermo Scientific Chemicals

Product Code. 11423850
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum tert-butoxide is utilized as a reagent in the Oppenauer oxidation of alcohols to ketones. It is also used in Meerwein-Ponndorf-Verley reduction of ketones. It is actively involved in the preparation of 1,1,2-triethoxyethene by reacting with 1,1,2,2-tetraethoxyethane followed by the elimination of alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-91-2
Molecular Formula C12H27AlO3-3
Molecular Weight (g/mol) 246.327
MDL Number MFCD00008801
InChI Key BXQSSDQHLWSBEX-UHFFFAOYSA-N
Synonym aluminum tert-butoxide, acmc-20alth, 2-propanol, 2-methyl-,aluminum salt 3:1
PubChem CID 57350944
IUPAC Name aluminum;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Al]

Specifications

Melting Point 200°C to 202°C
Density 1.0251 g/mL
Boiling Point 156°C (2 mmHg)
Odor Alcohol-like
Quantity 5 g
UN Number UN2925
Beilstein 4148229
Sensitivity Moisture sensitive
Merck Index 14,333
Solubility Information Very soluble in organic solvents like alcohols,benzene,toluene and xylene.
Formula Weight 246.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Aluminum tert-butoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'