Learn More
Heathrow Scientific™ Aluminum Test Tube Basket
Product Code. 12337569 Shop All Thermo Scientific CO₂ Incubators Products
Click to view available options
Dimensions (L x W x H):
152 x 152 x 152 mm
256 x 152 x 152 mm
332 x 228 x 178 mm
Unit Size:
Each
Product Code. 12337569
Brand: Heathrow Scientific™ HS20341C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Wash and store test tubes. Heathrow Scientific™ Aluminum Test Tube Basket can withstand repeated exposure to autoclaves.
  • Constructed of lightweight aluminum
  • Ideal for washing and storing test tubes
  • Withstands repeated autoclaving and use in the lab
Specifications
Autoclavable Autoclavable
Material Aluminum
For Use With (Application) Washing, Autoclaving, Storing Test Tubes
Dimensions (L x W x H) 332 x 228 x 178 mm