Learn More
Welch Ilmvac™ Hose Connector Adaptor
Designed for use with Welch Ilmvac™ rotary vane vacuum pumps.
Brand: Welch Ilmvac™ 701712
This item is not returnable.
View return policy
Description
- Stainless steel hose connector adaptor
- For 7mm tube
- DN 16 KF – DN 12
Specifications
Aluminum | |
1 Each | |
DN 16/12 | |
Transition Flange |
Product Content Correction
Product Title
Welch Ilmvac™ Hose Connector Adaptor > Size: DN 16/12
Spot an opportunity for improvement?Share a Content Correction