missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 0.81mm (0.032in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11359508
Click to view available options
Quantity:
10 m
250 m
50 m
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 50 m
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Weight ≈1.4g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Aluminum wire, 0.81mm (0.032 in.) dia., Annealed, Temper: soft

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'