missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Form: approx. 8.4g/m; Temper: soft

£60.70 - £822.09

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11335727
View Documents
Thermo Scientific Alfa Aesar
043428.G1
1 m
£60.70
1m
Please to purchase this item. Need a web account? Register with us today!
11345727
View Documents
Thermo Scientific Alfa Aesar
043428.G5
5 m
£217.00
5m
Please to purchase this item. Need a web account? Register with us today!
11325727
View Documents
Thermo Scientific Alfa Aesar
043428.G9
25 m
£822.09
25m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
[Al]
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629

Specifications

Silver
Silver-colored wire
99.9995% (Metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium
5359268
Odorless
Aluminum, Annealed
7429-90-5
2.7g/cm3
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
Wire
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-072-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only