Learn More
Tocris Bioscience™ AM 580
AM 580 synthesized to Ancillary Material Grade
Brand: Tocris Bioscience™ TB0760-RMU/10
Description
TB0760-RMU is an Ancillary Material Grade version of product AM 580 (Cat. No. 0760). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.Find out more about our Ancillary Material Grade product range.
Specifications
4-[[(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carbonyl]amino]benzoic acid | |
10 mg | |
C22H25NO3 |
102121-60-8 | |
Retinoic Acid Receptor Agonists | |
0.98 |