missing translation for 'onlineSavingsMsg'
Learn More

Faster™ AM Filter for ChemFAST 09 Top/Elite

Safety filter for the ChemFAST 09 Top/Elite.

Brand:  Faster™ FX0000844040

Code : NEW

Additional Details : Weight : 15.00000kg

Product Code. 15573336

  • £665.00 / Each
Estimated Shipment: 29-10-2024
to see stock.

Description

Description

  • Safety filter to be built into an internal Recirculating Fume Cupboard (dependent on chemical form)
  • Assists in filtering chemicals and vapor
  • Uses Ammonosorb Impregnated Activated Carbon
Specifications

Specifications

AMN
1.5 ft.
Activated Carbon
ChemFAST 09 Top/Elite
50 cm
Ammonia and Derivatives, Organic Emissions, Hydrocarbons, Aromatic Solvents
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers