Learn More
AM095 free acid, MedChemExpress
MedChemExpress AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.
£108.90 - £2162.27
Specifications
Chemical Name or Material | AM095 free acid |
---|---|
CAS | 1228690-36-5 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C27H24N2O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16473027
|
Medchem Express
HY-16040 |
50 mg |
£515.46
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16467046
|
Medchem Express
HY-16040 |
100 mg |
£801.02
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16459517
|
Medchem Express
HY-16040 |
5 mg |
£108.90
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16441216
|
Medchem Express
HY-16040 |
200 mg |
£1201.53
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411806
|
Medchem Express
HY-16040 |
500 mg |
£2162.27
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16406978
|
Medchem Express
HY-16040 |
10 mg |
£171.82
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.Specifications
AM095 free acid | |
Off-White | |
C27H24N2O5 | |
O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1 | |
456.49 | |
Research | |
COVID-19-immunoregulation |
1228690-36-5 | |
Research | |
DMSO : 67.3 mg/mL (147.43 mM; Need ultrasonic and warming) | |
456.49 | |
98.07% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.