Learn More
AM211, MedChemExpress
MedChemExpress AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.
£102.85 - £1132.56
Specifications
Chemical Name or Material | AM211 |
---|---|
CAS | 1175526-27-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C27H27F3N2O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498257
|
Medchem Express
HY-13213 |
5 mg |
£102.85
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16445558
|
Medchem Express
HY-13213 |
10 mg |
£171.82
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436116
|
Medchem Express
HY-13213 |
100 mg |
£1132.56
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16421727
|
Medchem Express
HY-13213 |
50 mg |
£675.18
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.Specifications
AM211 | |
White | |
C27H27F3N2O4 | |
DMSO : ≥ 125 mg/mL (249.75 mM) | |
500.51 | |
98.11% | |
Solid |
1175526-27-8 | |
Research | |
AM211 free acid | |
O=C(O)CC1=CC(C2=CC=C(C(F)(F)F)C=C2CN(CC)C(NCC3=CC=CC=C3)=O)=C(OC)C=C1 | |
500.51 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.