missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Amber Borosilicate Glass Class A Volumetric Flask with Stopper

Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.

Brand:  Fisherbrand™ FB55545

115.47 GBP valid until 2024-12-31
Use promo code "21985" to get your promotional price.


 View more versions of this product

Product Code. 11339443

  • £183.00 / Pack of 2
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.

With one mark graduation and interchangeable polyethylene stopper. The details on each flask include confirmation of compliance to International Standards as well as an individual re-order code. The Fisherbrand 'Batch Certificate of Compliance' is downloadable for this item from the 'Documentation' tab
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 1042, DIN 12664
Amber
Borosilicate Glass
±0.15 mL
A
Polyethylene
250 mL
220 mm
Yes
14/23
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers