Learn More
BRAND™ Amber Conical Ground Joint Stoppers
Boro 3.3 honed stoppers
Brand: BRAND™ 144671
Additional Details : Weight : 0.00350kg
Specifications
Honed conical joint stopper | |
NS 10/19 | |
1 | |
Borosilicate Glass |