missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hecht Karl™ Amber Glass Bottle with Conical Shoulder
Click to view available options
Capacity (Metric):
500 mL
Specifications
Specifications
Capacity (Metric) | 500 mL |
Color | Amber |
Material | Glass |
No. per Case | 1Each |
No. per Pack | 1Each |
Quantity | 1Each |
Neck Finish | Narrow Neck |
Nominal Capacity | 500mL |
Product Type | Reagent Bottle |
Closure Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hecht Karl™ Amber Glass Bottle with Conical Shoulder >
Spot an opportunity for improvement?Share a Content Correction