missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Amber Wide Neck Reagent Bottles
Click to view available options
Capacity (Metric):
100 mL
250 mL
1000 mL
Specifications
Capacity (Metric) | 250 mL |
Material | Soda Lime Glass |
Quantity | 10Pack |
Stopper Number | NS 34/24 |
Height (Metric) | 129 mm |
Product Type | Reagent Bottle |
Diameter (Metric) | 71 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'