missing translation for 'onlineSavingsMsg'
Learn More

Amberchrom™ 50WX8 50-100 (H)

Product Code. 11499617
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11499617

Brand: Thermo Scientific Alfa Aesar B22109.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11119-67-8

Dowex™ 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Spécification

Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX8 50-100 (H)
compliance-icons
Identifiant produit
  • Dowex 50WX8 50-100 (H)
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
Mention de danger
  • H318-Causes serious eye damage.
Conseils de prudence
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contains : Dowex 50WX8

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis