missing translation for 'onlineSavingsMsg'
Learn More

Amberchrom™ 50WX8 50-100 (H)

Product Code. p-7026222
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11489617

Brand: Thermo Scientific Alfa Aesar B22109.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11119-67-8

Dowex™ 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX8 50-100 (H)
compliance-icons
Product Identifier
  • Dowex 50WX8 50-100 (H)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dowex 50WX8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.